4-Chlor-3-fluor-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
022436-1G
FLUO022436-1GEA
1010
EUR
FLUO022436-1G
4-Chlor-3-fluor-3'-methylbenzhydrol
4-Chlor-3-fluor-3'-methylbenzhydrol
Formel:
C₁₄H₁₂ClFO |
MDL:
MFCD06201266 CAS-Nummer: 842140-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...