3-Chlor-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
022437-1G
FLUO022437-1GEA
1170
EUR
FLUO022437-1G
3-Chlor-3'-methylbenzhydrol
3-Chlor-3'-methylbenzhydrol
Formel:
C₁₄H₁₃ClO |
CAS-Nummer:
842140-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...