3,4-Dichlor-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022438-1G
FLUO022438-1GEA 1010 EUR
FLUO022438-1G
3,4-Dichlor-3'-methylbenzhydrol
3,4-Dichlor-3'-methylbenzhydrol
Formel: C₁₄H₁₂Cl₂O
MDL: MFCD06201268
CAS-Nummer: 842140-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR