3,5-Difluor-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
022440-1G
FLUO022440-1GEA
1010
EUR
FLUO022440-1G
3,5-Difluor-3'-methylbenzhydrol
3,5-Difluor-3'-methylbenzhydrol
Formel:
C₁₄H₁₂F₂O |
MDL:
MFCD06201270 CAS-Nummer: 842140-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...