3,4-Dimethoxy-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022441-1G
FLUO022441-1GEA 1010 EUR
FLUO022441-1G
3,4-Dimethoxy-3'-methylbenzhydrol
3,4-Dimethoxy-3'-methylbenzhydrol
Formel: C₁₆H₁₈O₃
CAS-Nummer: 844683-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR