4-(Dimethylamino)-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
022442-1G
FLUO022442-1GEA
1010
EUR
FLUO022442-1G
4-(Dimethylamino)-3'-methylbenzhydrol
4-(Dimethylamino)-3'-methylbenzhydrol
Formel:
C₁₆H₁₉NO |
CAS-Nummer:
844683-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...