4-(Ethylthio)-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022445-1G
FLUO022445-1GEA 1200 EUR
FLUO022445-1G
4-(Ethylthio)-3'-methylbenzhydrol
4-(Ethylthio)-3'-methylbenzhydrol
Formel: C₁₆H₁₈OS
MDL: MFCD06201274
CAS-Nummer: 844683-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR