3-Methoxy-3'-methylbenzhydrol
Lieferant: FLUOROCHEM
Synonyme:
3-Methoxy-α-(3-methylphenyl)benzenemethanol
022447-1G
FLUO022447-1GEA
1200
EUR
FLUO022447-1G
3-Methoxy-3'-methylbenzhydrol
3-Methoxy-3'-methylbenzhydrol
Formel:
C₁₅H₁₆O₂ Molecular Weight: 228,29 g/mol |
MDL:
MFCD06201276 CAS-Nummer: 844683-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...