4-Methoxy-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022448-1G
FLUO022448-1GEA 1010 EUR
FLUO022448-1G
4-Methoxy-3'-methylbenzhydrol
4-Methoxy-3'-methylbenzhydrol
Formel: C₁₅H₁₆O₂
MDL: MFCD06201277
CAS-Nummer: 838-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR