4-Chlor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022450-1G
FLUO022450-1GEA 461 EUR
FLUO022450-1G
4-Chlor-4'-methylbenzhydrol
4-Chlor-4'-methylbenzhydrol
Formel: C₁₄H₁₃ClO
MDL: MFCD06201279
CAS-Nummer: 13389-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR