3,4'-Dichlor-4-fluorbenzhydrol
Lieferant: FLUOROCHEM
022452-1G
FLUO022452-1GEA
1010
EUR
FLUO022452-1G
3,4'-Dichlor-4-fluorbenzhydrol
3,4'-Dichlor-4-fluorbenzhydrol
Formel:
C₁₃H₉Cl₂FO |
MDL:
MFCD06201281 CAS-Nummer: 844683-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...