3,4'-Dichlor-5-fluorbenzhydrol
Lieferant: FLUOROCHEM
022453-1G
FLUO022453-1GEA
1010
EUR
FLUO022453-1G
3,4'-Dichlor-5-fluorbenzhydrol
3,4'-Dichlor-5-fluorbenzhydrol
Formel:
C₁₃H₉Cl₂FO |
MDL:
MFCD06201282 CAS-Nummer: 844683-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...