4,4'-Dichlor-3-fluorbenzhydrol

Lieferant: FLUOROCHEM

022454-1G
FLUO022454-1GEA 1010 EUR
FLUO022454-1G
4,4'-Dichlor-3-fluorbenzhydrol
4,4'-Dichlor-3-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201283
CAS-Nummer: 844683-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR