3,4'-Dichlorbenzhydrol

Lieferant: FLUOROCHEM

022455-1G
FLUO022455-1GEA 344 EUR
FLUO022455-1G
3,4'-Dichlorbenzhydrol
3,4'-Dichlorbenzhydrol
Formel: C₁₃H₁₀Cl₂O
CAS-Nummer: 68240-78-8
EINECS: 269-481-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR