4-Chlor-3',4'-difluorbenzhydrol

Lieferant: FLUOROCHEM

022458-1G
FLUO022458-1GEA 1010 EUR
FLUO022458-1G
4-Chlor-3',4'-difluorbenzhydrol
4-Chlor-3',4'-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
CAS-Nummer: 844683-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR