4-Chlor-3',5'-difluorbenzhydrol

Lieferant: FLUOROCHEM

022459-1G
FLUO022459-1GEA 1010 EUR
FLUO022459-1G
4-Chlor-3',5'-difluorbenzhydrol
4-Chlor-3',5'-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
MDL: MFCD06201288
CAS-Nummer: 844683-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR