4-Chlor-3',4'-dimethoxybenzhydrol

Lieferant: FLUOROCHEM

022460-1G
FLUO022460-1GEA 1010 EUR
FLUO022460-1G
4-Chlor-3',4'-dimethoxybenzhydrol
4-Chlor-3',4'-dimethoxybenzhydrol
Formel: C₁₅H₁₅ClO₃
MDL: MFCD02127309
CAS-Nummer: 844683-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR