4-Chlor-4'-(dimethylamino)benzhydrol

Lieferant: FLUOROCHEM

022461-1G
FLUO022461-1GEA 1200 EUR
FLUO022461-1G
4-Chlor-4'-(dimethylamino)benzhydrol
4-Chlor-4'-(dimethylamino)benzhydrol
Formel: C₁₅H₁₆ClNO
MDL: MFCD06201289
CAS-Nummer: 844683-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR