4-Chlor-4'-(ethylthio)benzhydrol

Lieferant: FLUOROCHEM

022464-1G
FLUO022464-1GEA 1200 EUR
FLUO022464-1G
4-Chlor-4'-(ethylthio)benzhydrol
4-Chlor-4'-(ethylthio)benzhydrol
Formel: C₁₅H₁₅ClOS
MDL: MFCD06201292
CAS-Nummer: 844683-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR