4-Chlor-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022465-1G
FLUO022465-1GEA 143 EUR
FLUO022465-1G
4-Chlor-3'-fluorbenzhydrol
4-Chlor-3'-fluorbenzhydrol
Formel: C₁₃H₁₀ClFO
MDL: MFCD06201293
CAS-Nummer: 2795-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR