4-Chlor-4'-(methylthio)benzhydrol
Lieferant: FLUOROCHEM
022468-1G
FLUO022468-1GEA
1010
EUR
FLUO022468-1G
4-Chlor-4'-(methylthio)benzhydrol
4-Chlor-4'-(methylthio)benzhydrol
Formel:
C₁₄H₁₃ClOS |
MDL:
MFCD06201296 CAS-Nummer: 844683-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...