3-Chlor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

Synonyme: (3-Chlorophenyl)(p-tolyl)methanol

022469-1G
FLUO022469-1GEA 0 EUR
FLUO022469-1G
3-Chlor-4'-methylbenzhydrol
3-Chlor-4'-methylbenzhydrol
Formel: C₁₄H₁₃ClO
Molecular Weight: 232,710 g/mol
MDL: MFCD06201235
CAS-Nummer: 13395-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR