3,3'-Dichlor-4-fluorbenzhydrol

Lieferant: FLUOROCHEM

022471-1G
FLUO022471-1GEA 1010 EUR
FLUO022471-1G
3,3'-Dichlor-4-fluorbenzhydrol
3,3'-Dichlor-4-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201298
CAS-Nummer: 844683-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR