3,3'-Dichlor-5-fluorbenzhydrol

Lieferant: FLUOROCHEM

022472-1G
FLUO022472-1GEA 1010 EUR
FLUO022472-1G
3,3'-Dichlor-5-fluorbenzhydrol
3,3'-Dichlor-5-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201299
CAS-Nummer: 844683-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR