3,4'-Dichlor-3-fluorbenzhydrol

Lieferant: FLUOROCHEM

022473-1G
FLUO022473-1GEA 1010 EUR
FLUO022473-1G
3,4'-Dichlor-3-fluorbenzhydrol
3,4'-Dichlor-3-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201300
CAS-Nummer: 844683-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR