3,3'-Dichlorbenzhydrol
Lieferant: FLUOROCHEM
Synonyme:
Bis(3-chlorophenyl)methanol
022474-1G
FLUO022474-1GEA
104
EUR
FLUO022474-1G
3,3'-Dichlorbenzhydrol
3,3'-Dichlorbenzhydrol
Formel:
C₁₃H₁₀Cl₂O Molecular Weight: 253,123 g/mol |
MDL:
MFCD06201301 CAS-Nummer: 35190-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...