3-Chlor-3',4'-difluorbenzhydrol

Lieferant: FLUOROCHEM

022477-1G
FLUO022477-1GEA 1010 EUR
FLUO022477-1G
3-Chlor-3',4'-difluorbenzhydrol
3-Chlor-3',4'-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
MDL: MFCD06201304
CAS-Nummer: 844683-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR