3-Chlor-3',5'-difluorbenzhydrol

Lieferant: FLUOROCHEM

022478-1G
FLUO022478-1GEA 1010 EUR
FLUO022478-1G
3-Chlor-3',5'-difluorbenzhydrol
3-Chlor-3',5'-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
CAS-Nummer: 844683-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR