3-Chlor-3',4'-dimethoxybenzhydrol

Lieferant: FLUOROCHEM

022479-1G
FLUO022479-1GEA 1010 EUR
FLUO022479-1G
3-Chlor-3',4'-dimethoxybenzhydrol
3-Chlor-3',4'-dimethoxybenzhydrol
Formel: C₁₅H₁₅ClO₃
MDL: MFCD06201306
CAS-Nummer: 844683-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR