3-Chlor-3',5'-dimethylbenzhydrol

Lieferant: FLUOROCHEM

022481-1G
FLUO022481-1GEA 1010 EUR
FLUO022481-1G
3-Chlor-3',5'-dimethylbenzhydrol
3-Chlor-3',5'-dimethylbenzhydrol
Formel: C₁₅H₁₅ClO
MDL: MFCD06201308
CAS-Nummer: 844683-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR