3-Chlor-4'-(ethylthio)benzhydrol

Lieferant: FLUOROCHEM

022483-1G
FLUO022483-1GEA 1200 EUR
FLUO022483-1G
3-Chlor-4'-(ethylthio)benzhydrol
3-Chlor-4'-(ethylthio)benzhydrol
Formel: C₁₅H₁₅ClOS
MDL: MFCD06201310
CAS-Nummer: 844683-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR