3-Chlor-4'-(methylthio)benzhydrol
Lieferant: FLUOROCHEM
022487-1G
FLUO022487-1GEA
1010
EUR
FLUO022487-1G
3-Chlor-4'-(methylthio)benzhydrol
3-Chlor-4'-(methylthio)benzhydrol
Formel:
C₁₄H₁₃ClOS |
MDL:
MFCD06201314 CAS-Nummer: 844683-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...