4-tert-Butyl-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022488-1G
FLUO022488-1GEA 1010 EUR
FLUO022488-1G
4-tert-Butyl-3'-fluorbenzhydrol
4-tert-Butyl-3'-fluorbenzhydrol
Formel: C₁₇H₁₉FO
CAS-Nummer: 844683-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR