3-Chlor-3',4-difluorbenzhydrol

Lieferant: FLUOROCHEM

022489-1G
FLUO022489-1GEA 1010 EUR
FLUO022489-1G
3-Chlor-3',4-difluorbenzhydrol
3-Chlor-3',4-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
MDL: MFCD06201316
CAS-Nummer: 844683-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR