3-Chlor-3',5-difluorbenzhydrol

Lieferant: FLUOROCHEM

022490-1G
FLUO022490-1GEA 1010 EUR
FLUO022490-1G
3-Chlor-3',5-difluorbenzhydrol
3-Chlor-3',5-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
MDL: MFCD06201317
CAS-Nummer: 844683-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR