4-Chlor-3,3'-difluorbenzhydrol
Lieferant: FLUOROCHEM
022491-1G
FLUO022491-1GEA
1010
EUR
FLUO022491-1G
4-Chlor-3,3'-difluorbenzhydrol
4-Chlor-3,3'-difluorbenzhydrol
Formel:
C₁₃H₉ClF₂O |
MDL:
MFCD06201318 CAS-Nummer: 844683-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...