3,4-Dichlor-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022492-1G
FLUO022492-1GEA 1010 EUR
FLUO022492-1G
3,4-Dichlor-3'-fluorbenzhydrol
3,4-Dichlor-3'-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201319
CAS-Nummer: 844683-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR