3,5-Dichlor-3'-fluorbenzhydrol
Lieferant: FLUOROCHEM
022493-1G
FLUO022493-1GEA
1010
EUR
FLUO022493-1G
3,5-Dichlor-3'-fluorbenzhydrol
3,5-Dichlor-3'-fluorbenzhydrol
Formel:
C₁₃H₉Cl₂FO |
MDL:
MFCD06201320 CAS-Nummer: 844683-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...