3-Fluor-3',4'-dimethoxybenzhydrol

Lieferant: FLUOROCHEM

022496-1G
FLUO022496-1GEA 1010 EUR
FLUO022496-1G
3-Fluor-3',4'-dimethoxybenzhydrol
3-Fluor-3',4'-dimethoxybenzhydrol
Formel: C₁₅H₁₅FO₃
CAS-Nummer: 844683-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR