4-(Dimethylamino)-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022497-1G
FLUO022497-1GEA 1010 EUR
FLUO022497-1G
4-(Dimethylamino)-3'-fluorbenzhydrol
4-(Dimethylamino)-3'-fluorbenzhydrol
Formel: C₁₅H₁₆FNO
CAS-Nummer: 844683-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR