3,5-Dimethyl-3'-fluorbenzhydrol
Lieferant: FLUOROCHEM
022498-1G
FLUO022498-1GEA
1010
EUR
FLUO022498-1G
3,5-Dimethyl-3'-fluorbenzhydrol
3,5-Dimethyl-3'-fluorbenzhydrol
Formel:
C₁₅H₁₅FO |
CAS-Nummer:
844683-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...