4-Ethyl-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022499-1G
FLUO022499-1GEA 1200 EUR
FLUO022499-1G
4-Ethyl-3'-fluorbenzhydrol
4-Ethyl-3'-fluorbenzhydrol
Formel: C₁₅H₁₅FO
MDL: MFCD06201326
CAS-Nummer: 844683-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR