4-(Ethylthio)-3'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022500-1G
FLUO022500-1GEA 1200 EUR
FLUO022500-1G
4-(Ethylthio)-3'-fluorbenzhydrol
4-(Ethylthio)-3'-fluorbenzhydrol
Formel: C₁₅H₁₅FOS
MDL: MFCD06201327
CAS-Nummer: 844683-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR