3,3'-Difluorbenzhydrol

Lieferant: FLUOROCHEM

022501-1G
FLUO022501-1GEA 209 EUR
FLUO022501-1G
3,3'-Difluorbenzhydrol
3,3'-Difluorbenzhydrol
Formel: C₁₃H₁₀F₂O
MDL: MFCD03077396
CAS-Nummer: 98586-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR