4-Fluor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022504-1G
FLUO022504-1GEA 1010 EUR
FLUO022504-1G
4-Fluor-4'-methylbenzhydrol
4-Fluor-4'-methylbenzhydrol
Formel: C₁₄H₁₃FO
CAS-Nummer: 345-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR