4-tert-Butyl-4'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022505-1G
FLUO022505-1GEA 321 EUR
FLUO022505-1G
4-tert-Butyl-4'-fluorbenzhydrol
4-tert-Butyl-4'-fluorbenzhydrol
Formel: C₁₇H₁₉FO
CAS-Nummer: 596812-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR