3-Chlor-4,4'-difluorbenzhydrol
Lieferant: FLUOROCHEM
022506-1G
FLUO022506-1GEA
1010
EUR
FLUO022506-1G
3-Chlor-4,4'-difluorbenzhydrol
3-Chlor-4,4'-difluorbenzhydrol
Formel:
C₁₃H₉ClF₂O |
CAS-Nummer:
191591-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...