4-Chlor-3,4'-difluorbenzhydrol

Lieferant: FLUOROCHEM

022508-1G
FLUO022508-1GEA 702 EUR
FLUO022508-1G
4-Chlor-3,4'-difluorbenzhydrol
4-Chlor-3,4'-difluorbenzhydrol
Formel: C₁₃H₉ClF₂O
CAS-Nummer: 844683-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR