3-Chlor-4'-fluorbenzhydrol
Lieferant: FLUOROCHEM
022509-1G
FLUO022509-1GEA
531
EUR
FLUO022509-1G
3-Chlor-4'-fluorbenzhydrol
3-Chlor-4'-fluorbenzhydrol
Formel:
C₁₃H₁₀ClFO |
MDL:
MFCD06201335 CAS-Nummer: 97759-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...