3,4-Dichlor-4'-fluorbenzhydrol

Lieferant: FLUOROCHEM

022510-1G
FLUO022510-1GEA 1010 EUR
FLUO022510-1G
3,4-Dichlor-4'-fluorbenzhydrol
3,4-Dichlor-4'-fluorbenzhydrol
Formel: C₁₃H₉Cl₂FO
MDL: MFCD06201336
CAS-Nummer: 211293-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR